Thiourea, 99%, Thermo Scientific Chemicals
Thiourea, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiourea, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
10,000 g
Catalog number A12828.0E
also known as A12828-0E
Price (USD)/ Each
131.65
Online exclusive
155.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
131.65
Online exclusive
155.00 
Save 23.35 (15%)
Add to cart
Thiourea, 99%, Thermo Scientific Chemicals
Catalog numberA12828.0E
Price (USD)/ Each
131.65
Online exclusive
155.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
CAS62-56-6
IUPAC Namethiourea
Molecular FormulaCH4N2S
InChI KeyUMGDCJDMYOKAJW-UHFFFAOYSA-N
SMILESNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder
Elemental AnalysisSulfur content 41.49-42.75% (dry weight basis)
Loss on Drying≤1.0% (@105?C to constant weight)
Assay (unspecified)(by Sulfur EA) : ≥ 98.5 to ≤ 101.5% (dry weight basis)
View more
Reagent for organic synthesisUsed as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It acts as a catalyst for asymmetric reactions. It plays an essential role as a catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for organic synthesisUsed as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It acts as a catalyst for asymmetric reactions. It plays an essential role as a catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.

Solubility
Soluble in water.

Notes
Stable. Incompatible with strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Reagent for the conversion of alkyl halides to thiols by base hydrolysis of the isothiouronium salts; see also N-Acetyl thiourea, B21198. Cleavage of isothiouronium salts with base and alkylation of the resulting thiolate has been used as a convenient synthesis of unsymmetrical sulfides: Synth. Commun., 14, 209 (1984).
  2. Epoxides are converted to episulfides: J. Org. Chem., 26, 3467 (1961). The 2,3-epoxy alcohols resulting from the Sharpless enantioselective epoxidation can be converted to the corresponding episulfides with retention at both centers, using Ti(O-i-Pr)4 as mediator: J. Org. Chem., 53, 4114 (1988).
  3. Widely used in heterocyclic syntheses, e.g. of thiazoles and pyrimidines.
  4. Has been used in a convenient synthesis of isothiocyanates from oximes via the nitrile oxide: Tetrahedron Lett., 34, 8283 (1993); see also Benzaldoxime, A12053:
  5. Bui, T.; Syed, S.; Barbas, C. F. Thiourea-Catalyzed Highly Enantio- and Diastereoselective Additions of Oxindoles to Nitroolefins: Application to the Formal Synthesis of (+) -Physostigmine. J. Am. Chem. Soc. 2009, 131 (25), 8758-8759.
  6. Takemoto, Y. Development of Chiral Thiourea Catalysts and Its Application to Asymmetric Catalytic Reactions. Chem. Pharm. Bull. 2010, 58 (5), 593-601.