Sodium 1-dodecanesulfonate, 99%, Thermo Scientific Chemicals
Sodium 1-dodecanesulfonate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium 1-dodecanesulfonate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
10 g
Catalog number A12829.14
also known as A12829-14
Price (USD)/ Each
346.65
Online exclusive
385.00 
Save 38.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
346.65
Online exclusive
385.00 
Save 38.35 (10%)
Add to cart
Sodium 1-dodecanesulfonate, 99%, Thermo Scientific Chemicals
Catalog numberA12829.14
Price (USD)/ Each
346.65
Online exclusive
385.00 
Save 38.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2386-53-0
IUPAC Namesodium dodecane-1-sulfonate
Molecular FormulaC12H25NaO3S
InChI KeyDAJSVUQLFFJUSX-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥98.5%
Water Content (Karl Fischer Titration)≤1.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (unspecified)≥98.5%
View more
Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Jin, Y.; Wang, L.; Wang, T.; Chen, P.; Bi, Y.; Yu, L. Aggregation behavior of dodecylsulfonate-based surface active ionic liquids in water. J. Mol. Liq. 2015, 212, 23-29.
  2. Khan, A.; Khan, A. A. P; Asiri, A. M.; Alamry, K. A. Preparation and Characterization of hybrid graphene oxide composite and its application in paracetamol microbiosensor. Polym. Compos. 2015, 36, (2), 221-228.