Thermo Scientific Chemicals

Ammonium copper(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: A12834.0B
1000 g, Each
Thermo Scientific Chemicals

Ammonium copper(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: A12834.0B
1000 g, Each
Quantity
Catalog number: A12834.0B
also known as A12834-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13587-26-3
IUPAC Name
copper(2+) diamine hexahydrate dihydrogen sulfate
Molecular Formula
CuH20N2O14S2
InChI Key
IHEZHABINDDXAE-UHFFFAOYSA-L
SMILES
N.N.O.O.O.O.O.O.[Cu++].OS([O-])(=O)=O.OS([O-])(=O)=O
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
Blue
Assay (Iodometric Titration)
≥98.5 to ≤101.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text