Thermo Scientific Chemicals

Pyrrole-2-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A12838.14
25 g, Each
Thermo Scientific Chemicals

Pyrrole-2-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A12838.14
25 g, Each
Quantity
Catalog number: A12838.14
also known as A12838-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1003-29-8
IUPAC Name
1H-pyrrole-2-carbaldehyde
Molecular Formula
C5H5NO
InChI Key
ZSKGQVFRTSEPJT-UHFFFAOYSA-N
SMILES
O=CC1=CC=CN1
Free acid (titration)
<0.5%
Form
Crystalline powder, crystals or fused solid
Assay (GC)
≥98.5%
Appearance (Color)
White to cream or brown
Melting Point (clear melt)
40-47?C

Description

Pyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.

Solubility
Soluble in chloroform, dimethyl sulfoxide and methanol. Insoluble in water.

Notes
Store in cool place. Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text