Thermo Scientific Chemicals

2,4,6-Trichloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: A12842.14
25 g, Each
Thermo Scientific Chemicals

2,4,6-Trichloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: A12842.14
25 g, Each
Quantity
Catalog number: A12842.14
also known as A12842-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3764-01-0
IUPAC Name
2,4,6-trichloropyrimidine
Molecular Formula
C4HCl3N2
InChI Key
DPVIABCMTHHTGB-UHFFFAOYSA-N
SMILES
ClC1=CC(Cl)=NC(Cl)=N1
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5675-1.5715 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

It is applied In selective Suzuki coupling with arylboronic acids has been reported to give 6-arylpyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied In selective Suzuki coupling with arylboronic acids has been reported to give 6-arylpyrimidines.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text