1-Benzyl-3-piperidone hydrochloride hydrate, Thermo Scientific Chemicals
1-Benzyl-3-piperidone hydrochloride hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Benzyl-3-piperidone hydrochloride hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A12849.14
also known as A12849-14
Price (USD)/ Each
512.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
512.00
Add to cart
1-Benzyl-3-piperidone hydrochloride hydrate, Thermo Scientific Chemicals
Catalog numberA12849.14
Price (USD)/ Each
512.00
-
Add to cart
Chemical Identifiers
CAS50606-58-1
IUPAC Name1-benzylpiperidin-3-one hydrochloride
Molecular FormulaC12H16ClNO
InChI KeyOVWSFXNSJDMRPV-UHFFFAOYSA-N
SMILESCl.O=C1CCCN(CC2=CC=CC=C2)C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Cream to pale brown
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Water Content (Karl Fischer Titration)≤8.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only