Thermo Scientific Chemicals

Behenic acid, tech. 85%, Thermo Scientific Chemicals

Catalog number: A12850.0I
5000 g, Each
Thermo Scientific Chemicals

Behenic acid, tech. 85%, Thermo Scientific Chemicals

Catalog number: A12850.0I
5000 g, Each
Quantity
Catalog number: A12850.0I
also known as A12850-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-85-6
IUPAC Name
docosanoic acid
Molecular Formula
C22H44O2
InChI Key
UKMSUNONTOPOIO-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCC(O)=O
Appearance (Color)
White to pale cream or pale yellow
Assay (Silylated GC)
≥83.0%
Comment
C24 = 0 - 3%: C26 = 0 - 0.5%
Melting Point (clear melt)
74-82?C
Identification (FTIR)
Conforms

Description

Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.

Solubility
Soluble in DMF (∼3 mg/ml), hot methanol, water (0.15 mg/ml at 25°C), chloroform, and ethanol (2.18 mg/ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, reducing agents and bases. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text