Gibco™

CTS™ DPBS, without calcium chloride, without magnesium chloride

產品號碼: A1285602
Gibco™

CTS™ DPBS, without calcium chloride, without magnesium chloride

產品號碼: A1285602
產品號碼
A1285602
包裝規格
2 L
價格 (HKD)
價格 3,104.00
您的價格
供應狀況
***
數量
產品號碼
包裝規格
1 L
價格 (HKD)
價格 579.00
您的價格
供應狀況
***
數量
產品號碼包裝規格價格 (HKD)供應狀況數量
A12856022 L
價格 3,104.00
您的價格
***
A12856011 L
價格 579.00
您的價格
***
產品概覽
圖表
影片
建議
建議
文件
常見問題
引用資料與參考文獻
其他資訊
建議

Gibco Cell Therapy Systems (CTS) DPBS is the classic Dulbecco's phosphate-buffered saline. This particular formulation lacks calcium, magnesium, and phenol red.

CTS DPBS is manufactured in cGMP, ISO-certified facilities to help ensure the quality and consistency for reproducible results every day. Tests include osmolality, pH, stability, and the absence of bacterial/fungal contamination and endotoxin.

Gibco CTS products are high quality and supplied with harmonized documentation such as certificates of analysis, certificates of origin, and access to authorization letters for our Drug Master Files, as appropriate. The CTS product line enables you to reduce your burden when qualifying reagents during your transition from research to clinical applications.

Closed systems compatible
This CTS DPBS is provided in bioprocess container format with a chamber constructed from Aegis 5-14 film. The sterile, weldable DEPH-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector provide versatile connection options for closed system manufacturing processes.

規格

Additive
No Calcium, No Magnesium
Classification
AOF
Dilution
1X
Product Type
DPBS
Quantity
2 L
Solution Type
Dulbecco's Phosphate Buffered Saline
For Use With (Application)
Cell and Gene Therapy Research, Development, and Manufacturing
Green Features
Sustainable packaging
Shelf Life
36 months from date of manufacture
Shipping Condition
Room Temperature
Sterility
Sterile-filtered
Form
Liquid
Format
Bag
Manufacturing Quality
ISO13485, MDSAP, FDA-registered, 21 CFR820

內容物與存放

Store at 15–30°C.

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text