Thermo Scientific Chemicals

4-(Chloromethyl)pyridine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12859.14
25 g, Each
Thermo Scientific Chemicals

4-(Chloromethyl)pyridine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12859.14
25 g, Each
Quantity
Catalog number: A12859.14
also known as A12859-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1822-51-1
IUPAC Name
hydrogen 4-(chloromethyl)pyridine chloride
Molecular Formula
C6H7Cl2N
InChI Key
ZDHKVKPZQKYREU-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClCC1=CC=NC=C1
Form
Powder
Water Content (Karl Fischer Titration)
≤1%
Appearance (Color)
Pale cream to cream to yellow to pale brown
Melting Point (clear melt)
163.0-170.0?C
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

It finds its application as a reagent for protection of carboxyl termini of peptides as 4-picolyl esters, providing a polar 'handle' which aids in separation and purification of the peptide. The group can be introduced in the presence of a base, e.g. tetramethylguanidine, is stable to acid-catalyzed removal of Cbz protecting group and can be removed by base, Na in liquid ammonia or catalytic hydrogenolysis. It is also applied in conjunction with Amberlyst™15 resin in peptide synthesis. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a reagent for protection of carboxyl termini of peptides as 4-picolyl esters, providing a polar ′handle′ which aids in separation and purification of the peptide. The group can be introduced in the presence of a base, e.g. tetramethylguanidine, is stable to acid-catalyzed removal of Cbz protecting group and can be removed by base, Na in liquid ammonia or catalytic hydrogenolysis. It is also applied in conjunction with Amberlyst™15 resin in peptide synthesis. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water (750 mg/ml at 20°C), soluble in 1N HCl (2.5%), DMSO, and methanol.

Notes
Hygroscopic. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text