Thermo Scientific Chemicals

2,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A12868.14
25 g, Each
Thermo Scientific Chemicals

2,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A12868.14
25 g, Each
Quantity
Catalog number: A12868.14
also known as A12868-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-01-2
IUPAC Name
2,4-dihydroxybenzaldehyde
Molecular Formula
C7H6O3
InChI Key
IUNJCFABHJZSKB-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=O)C(O)=C1
Identification (FTIR)
Conforms
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
132.0-139.0?C
Appearance (Color)
Pale cream to pale pink to pale brown
Form
Powder

Description

2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.

Solubility
Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol, ethyl ether, chloroform, glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text