Gibco™

Water For Injection (WFI) for Cell Culture

Catalog number: A1287302
Have Questions?
Gibco™

Water For Injection (WFI) for Cell Culture

Catalog number: A1287302
Have Questions?
Catalog Number
A1287302
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
A1287304
also known as A12873-04
Unit Size
6 x 1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A12873021,000 mL
-
A1287301500 mL
-
A128730310 x 500 mL
-
A1287304
also known as A12873-04
6 x 1 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco™ Water for Injection (WFI) for Cell Culture is high quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, Gibco™ Water for Injection (WFI) for Cell Culture features:
• Water For Injection Quality
• Gibco™ quality and performance
• Use in cell culture and molecular biology applications

Water for injection quality
Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere. The water is prepared by distillation and filtration (0.1 μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly.

Gibco™ quality and performance
The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Water for Injection (WFI) for Cell Culture. In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances.

Use in cell culture and molecular biology applications
Gibco™ Water for Injection (WFI) for Cell Culture can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions and diluting buffers.

Product Use
For Research Use Only or Further Manufacturing. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System
Gibco™ Water for Injection (WFI) for Cell Culture is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

pH
4 to 6
Recommended Storage
Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Grade
Cell Culture
Packaging
Bottle
Sterility
Sterile-filtered
Treatment(s)
None
Quantity
1000 mL
Purification Method
Distilled and filtered (0.1 μm)
Shipping Condition
Room Temperature
Product Line
Gibco™
Green Features
Sustainable packaging

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text