Thermo Scientific Chemicals

Diisopropyl ketone, 98%, Thermo Scientific Chemicals

Catalog number: A12875.22
100 g, Each
Thermo Scientific Chemicals

Diisopropyl ketone, 98%, Thermo Scientific Chemicals

Catalog number: A12875.22
100 g, Each
Quantity
Catalog number: A12875.22
also known as A12875-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
565-80-0
IUPAC Name
2,4-dimethylpentan-3-one
Molecular Formula
C7H14O
InChI Key
HXVNBWAKAOHACI-UHFFFAOYSA-N
SMILES
CC(C)C(=O)C(C)C
Form
Liquid
Refractive Index
1.3980-1.4020 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text