Thermo Scientific Chemicals

Phenylselenol, 90+%, Thermo Scientific Chemicals

Catalog number: A12876.14
25 g, Each
Thermo Scientific Chemicals

Phenylselenol, 90+%, Thermo Scientific Chemicals

Catalog number: A12876.14
25 g, Each
Quantity
Catalog number: A12876.14
also known as A12876-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
645-96-5
IUPAC Name
phenylsulfanyl
Molecular Formula
C6H5Se
InChI Key
YWBHROUQJYHSOR-UHFFFAOYSA-N
SMILES
[Se]C1=CC=CC=C1
Form
Liquid
Assay (GC)
≥90.0%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.6110-1.6190 @ 20?C

Description

It is applied for reactions of phenylselenol and its anion. Benzeneselenol can used as starting reagent in the synthesis of monoseleno-substituted 1,3-dienes. It may be used in the synthesis of cationic chalcogenolato-bridged diruthenium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied for reactions of phenylselenol and its anion. Benzeneselenol can used as starting reagent in the synthesis of monoseleno-substituted 1,3-dienes. It may be used in the synthesis of cationic chalcogenolato-bridged diruthenium complexes.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text