Thermo Scientific Chemicals

2-Methylcyclohexanol, cis + trans, 97%, Thermo Scientific Chemicals

Catalog number: A12877.AP
500 mL, Each
Thermo Scientific Chemicals

2-Methylcyclohexanol, cis + trans, 97%, Thermo Scientific Chemicals

Catalog number: A12877.AP
500 mL, Each
Quantity
Catalog number: A12877.AP
also known as A12877-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
583-59-5
IUPAC Name
2-methylcyclohexan-1-ol
Molecular Formula
C7H14O
InChI Key
NDVWOBYBJYUSMF-UHFFFAOYNA-N
SMILES
CC1CCCCC1O
Refractive Index
1.4615-1.4665 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Form
Liquid
Identification (FTIR)
Conforms

Description

2-Methylcyclohexanol is used in the preparation of acetic acid-(2-methyl-cyclohexyl ester) by reaction with acetic anhydride. Further, it is used to study the effect of organic solvents on epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanol is used in the preparation of acetic acid-(2-methyl-cyclohexyl ester) by reaction with acetic anhydride. Further, it is used to study the effect of organic solvents on epoxide hydrolase.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text