Thermo Scientific Chemicals

Cerium(IV) ammonium nitrate, 98+%, Thermo Scientific Chemicals

Catalog number: A12882.0B
1000 g, Each
Thermo Scientific Chemicals

Cerium(IV) ammonium nitrate, 98+%, Thermo Scientific Chemicals

Catalog number: A12882.0B
1000 g, Each
Quantity
Catalog number: A12882.0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16774-21-3
IUPAC Name
λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
Molecular Formula
CeH8N8O18
InChI Key
WIBGOERAEYJBOT-UHFFFAOYSA-N
SMILES
N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Assay (unspecified)
>98.0%
Form
Crystals or Crystalline powder
Appearance (Color)
Orange
Residual water
≤2.0%
Total rare earth oxides
ca 30.0%

Description

Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

Solubility
Soluble in water and alcohol.

Notes
Store in cool place. Hygroscopic. Incompatible with strong reducing agents and powdered metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text