Thermo Scientific Chemicals

Tris[4-(trifluoromethyl)phenyl]phosphine, 98%, Thermo Scientific Chemicals

Catalog number: A12885.03
1 g, Each
Thermo Scientific Chemicals

Tris[4-(trifluoromethyl)phenyl]phosphine, 98%, Thermo Scientific Chemicals

Catalog number: A12885.03
1 g, Each
Quantity
Catalog number: A12885.03
also known as A12885-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13406-29-6
IUPAC Name
tris[4-(trifluoromethyl)phenyl]phosphane
Molecular Formula
C21H12F9P
InChI Key
PXYCJKZSCDFXLR-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(C=C1)P(C1=CC=C(C=C1)C(F)(F)F)C1=CC=C(C=C1)C(F)(F)F
Appearance (Color)
White to yellow
Form
Crystals or Crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
72-76°C

Description

Tris[4-(trifluoromethyl)phenyl]phosphine is a raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris[4-(trifluoromethyl)phenyl]phosphine is a raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text