Thermo Scientific Chemicals

(Ethoxycarbonylmethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals

Catalog number: A12896.30
250 g, Each
Thermo Scientific Chemicals

(Ethoxycarbonylmethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals

Catalog number: A12896.30
250 g, Each
Quantity
Catalog number: A12896.30
also known as A12896-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1099-45-2
IUPAC Name
ethyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Molecular Formula
C22H21O2P
InChI Key
IIHPVYJPDKJYOU-UHFFFAOYSA-N
SMILES
CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Assay (HPLC)
≥98.0%
Appearance (Color)
White to pale cream or pale yellow
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

(Ethoxycarbonylmethylene)triphenylphosphoran is used as a Wittig reagent in organic synthesis. It is used in the preparation of indole from o-nitrobenzaldehydes, coumarins from o-hydroxy acetophenone. It acts as an inhibitor and inhibits the activity of cholinesterase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Ethoxycarbonylmethylene)triphenylphosphoran is used as a Wittig reagent in organic synthesis. It is used in the preparation of indole from o-nitrobenzaldehydes, coumarins from o-hydroxy acetophenone. It acts as an inhibitor and inhibits the activity of cholinesterase.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text