Thermo Scientific Chemicals

Iodotrimethylsilane, 97%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A12902.06
5 g, Each
Thermo Scientific Chemicals

Iodotrimethylsilane, 97%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A12902.06
5 g, Each
Quantity
Catalog number: A12902.06
also known as A12902-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16029-98-4
IUPAC Name
iodotrimethylsilane
Molecular Formula
C3H9ISi
InChI Key
CSRZQMIRAZTJOY-UHFFFAOYSA-N
SMILES
C[Si](C)(C)I
Appearance (Color)
Clear colorless to red or brown
Form
Liquid
Assay (unspecified)
≥96.0%
Comment
Material Sourced in the U.S. and in other countries
Comment
Stabilised with Copper powder. Assay performed by Titration or GC.

Description

Iodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.

Solubility
Miscible with water.

Notes
Moisture and light Sensitive. Incompatible withstrong acids, strong bases, strong oxidizing agents and reacts violently with water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text