Thermo Scientific Chemicals

Diisopropyl methylphosphonate, 95%, Thermo Scientific Chemicals

Catalog number: A12904.14
25 g, Each
Thermo Scientific Chemicals

Diisopropyl methylphosphonate, 95%, Thermo Scientific Chemicals

Catalog number: A12904.14
25 g, Each
Quantity
Catalog number: A12904.14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1445-75-6
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms
Refractive Index
1.4070-1.4130 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

Diisopropyl methylphosphonate is used as a precursor to prepare beta-keto phosphonates. It acts as an intermediate for Horner-Wadsworth-Emmons olefination of carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropyl methylphosphonate is used as a precursor to prepare beta-keto phosphonates. It acts as an intermediate for Horner-Wadsworth-Emmons olefination of carbonyl compounds.

Solubility
Miscible with water.

Notes
Store in cool place. Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, reducing agents, strong reducing agents and phosphorus halides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text