Thermo Scientific Chemicals

Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A12905.30
250 g, Each
Thermo Scientific Chemicals

Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A12905.30
250 g, Each
Quantity
Catalog number: A12905.30
also known as A12905-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10326-27-9
IUPAC Name
barium(2+) dihydrate dichloride
Molecular Formula
BaCl2H4O2
InChI Key
PWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Ba++]
Appearance (Color)
White to pale grey
Assay (Titration ex Chloride)
≥99.0 to ≤101.0% (non-U.S. specification)
Assay from Supplier's CofA
≥99.0% (U.S. specification)
Solution Test
(10% in water) : Clear & colorless (non-U.S. specification)
Form
Crystalline powder

Description

Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

Solubility
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.

Notes
Stable and hygroscopic in nature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text