Thermo Scientific Chemicals

9-Fluorenone, 98+%, Thermo Scientific Chemicals

Catalog number: A12910.22
100 g, Each
Thermo Scientific Chemicals

9-Fluorenone, 98+%, Thermo Scientific Chemicals

Catalog number: A12910.22
100 g, Each
Quantity
Catalog number: A12910.22
also known as A12910-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
486-25-9
IUPAC Name
9H-fluoren-9-one
Molecular Formula
C13H8O
InChI Key
YLQWCDOCJODRMT-UHFFFAOYSA-N
SMILES
O=C1C2=CC=CC=C2C2=CC=CC=C12
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
80.0-86.0?C
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder or flakes

Description

9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.

Solubility
Soluble in water, alcohol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text