Thermo Scientific Chemicals

MOPS, 99%, Thermo Scientific Chemicals

Catalog number: A12914.14
25 g, Each
Thermo Scientific Chemicals

MOPS, 99%, Thermo Scientific Chemicals

Catalog number: A12914.14
25 g, Each
Quantity
Catalog number: A12914.14
also known as A12914-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1132-61-2
IUPAC Name
4-(3-sulfonatopropyl)morpholin-4-ium
Molecular Formula
C7H15NO4S
InChI Key
DVLFYONBTKHTER-UHFFFAOYSA-N
SMILES
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%

Description

MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

Solubility
Soluble in water.    

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text