Thermo Scientific Chemicals

1-Acetylnaphthalene, 97+%, Thermo Scientific Chemicals

Catalog number: A12918.0B
1000 g, Each
Thermo Scientific Chemicals

1-Acetylnaphthalene, 97+%, Thermo Scientific Chemicals

Catalog number: A12918.0B
1000 g, Each
Quantity
Catalog number: A12918.0B
also known as A12918-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
941-98-0
IUPAC Name
1-(naphthalen-1-yl)ethan-1-one
Molecular Formula
C12H10O
InChI Key
QQLIGMASAVJVON-UHFFFAOYSA-N
SMILES
CC(=O)C1=C2C=CC=CC2=CC=C1
Refractive Index
1.6250-1.6300 @ 20?C
Appearance (Color)
Clear colorless to yellow to orange
Assay (GC)
≥97.0%
Form
Liquid

Description

They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Not miscible or difficult to mix in water. Soluble in ethyl alcohol. Insoluble in paraffin oil.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text