Thermo Scientific Chemicals

L-Glutamic acid monosodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A12919.0B
1000 g, Each
Thermo Scientific Chemicals

L-Glutamic acid monosodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A12919.0B
1000 g, Each
Quantity
Catalog number: A12919.0B
also known as A12919-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6106-04-3
IUPAC Name
sodium (4S)-4-amino-4-carboxybutanoate hydrate
Molecular Formula
C5H10NNaO5
InChI Key
GJBHGUUFMNITCI-QTNFYWBSSA-M
SMILES
O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O
Form
Crystalline powder
Assay from Supplier's CofA
≥98.0% (U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)
Comment
Purchased in the U.S. and in other countries
Appearance (Color)
White to pale cream

Description

L-Glutamic acid monosodium salt monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. Also used to intensify flavors of foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid monosodium salt monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. Also used to intensify flavors of foods.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text