Thermo Scientific Chemicals

2-Bromopropane, 99%, Thermo Scientific Chemicals

Catalog number: A12944.AK
250 mL, Each
Thermo Scientific Chemicals

2-Bromopropane, 99%, Thermo Scientific Chemicals

Catalog number: A12944.AK
250 mL, Each
Quantity
Catalog number: A12944.AK
also known as A12944-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-26-3
IUPAC Name
2-bromopropane
Molecular Formula
C3H7Br
InChI Key
NAMYKGVDVNBCFQ-UHFFFAOYSA-N
SMILES
CC(C)Br
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4235-1.4265 @ 20?C

Description

2-Bromopropane serves as an alkylating agent in organic synthesis. It is also used as an intermediate to form alkylated amines and alkylated metallic compounds. Further, it acts as a solvent for industrial cleaning, degreasing, metal processing and finishing, electronics, aerospace and aviation, aerosols, textiles, adhesives and inks. In addition, it is used for introducing the isopropyl functional group in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropane serves as an alkylating agent in organic synthesis. It is also used as an intermediate to form alkylated amines and alkylated metallic compounds. Further, it acts as a solvent for industrial cleaning, degreasing, metal processing and finishing, electronics, aerospace and aviation, aerosols, textiles, adhesives and inks. In addition, it is used for introducing the isopropyl functional group in organic synthesis.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water and acetone.

Notes
Avoid all possible sources of ignition.Incompatible with strong oxidizing agents and strong bases.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text