Thermo Scientific Chemicals

4-Fluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A12946.22
100 g, Each
Thermo Scientific Chemicals

4-Fluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A12946.22
100 g, Each
Quantity
Catalog number: A12946.22
also known as A12946-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
403-43-0
IUPAC Name
4-fluorobenzoyl chloride
Molecular Formula
C7H4ClFO
InChI Key
CZKLEJHVLCMVQR-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)C(Cl)=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.5295-1.5335 @ 20?C

Description

It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text