It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under gas.
RUO – Research Use Only
Ohno M, et al. Synthesis of a novel naphthalene-based poly (arylene ether-ketone) by polycondensation of 1, 5-bis (4-fluorobenzoyl)-2, 6-dimethylnaphthalene with bisphenol A.J. Polym. Sci. A Polym. Chem.,1995,33(15), 2647-2655.
Frank W. Mercer, et al. Synthesis and characterization of fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties.J. Polym. Sci., Part A: Polym. Chem.,1997,35(3), 521-526.