Biological stain for distinguishing coli and aerogene bacteriaBasic Fuchsin is useful for Ziehl-Neelson staining to detect acid-fast bacilli. It is also used to stain tubercle bacillus and bacterial flagella. It plays an important role for tissue sections to visualize elastic fibers, cardiac and skeletal muscle tissue. It also acts as bacterial and histological stains. Further, it is used to stain cellular elements, certain tissues, bacteria and also acts as a local anti-infective.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Biological stain for distinguishing coli and aerogene bacteriaBasic Fuchsin is useful for Ziehl-Neelson staining to detect acid-fast bacilli. It is also used to stain tubercle bacillus and bacterial flagella. It plays an important role for tissue sections to visualize elastic fibers, cardiac and skeletal muscle tissue. It also acts as bacterial and histological stains. Further, it is used to stain cellular elements, certain tissues, bacteria and also acts as a local anti-infective.
Solubility
Soluble in water, alcohol, diethyl ether and acids practice. Insoluble in ether.
Notes
Incompatible with oxidizing agents, reducing agents, acids, acid chlorides, acid anhydrides and chloroformates.
RUO – Research Use Only
Yang, X.; Li , Y.; Du, Q.; Sun, J.; Chen, L.; Hu, S.; Wang, Z.; Xia, Y.; Xia, L. Highly effective removal of basic fuchsin from aqueous solutions by anionic polyacrylamide/graphene oxide aerogels. J. Colloid Interface Sci. 2015, 453, 107-114.
Kumawat, P.; Joshi, M.; Ameta, R.; Ameta, S. C. Photocatalytic degradation of basic Fuchsin over quaternary oxide iron zinc cuprate (FeZn2cu3O6.5). Adv. App. Sci. Res. 2015, 6 (7), 209-215.