n-Nonylbenzene, 97%, Thermo Scientific Chemicals
n-Nonylbenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Nonylbenzene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A12957.18
also known as A12957-18
Price (USD)/ Each
139.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
139.00
Add to cart
n-Nonylbenzene, 97%, Thermo Scientific Chemicals
Catalog numberA12957.18
Price (USD)/ Each
139.00
-
Add to cart
Chemical Identifiers
CAS1081-77-2
IUPAC Namenonylbenzene
Molecular FormulaC15H24
InChI KeyLIXVMPBOGDCSRM-UHFFFAOYSA-N
SMILESCCCCCCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4815-1.4865 @ 20?C
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only