Thermo Scientific Chemicals

Adamantane-1-carboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12959.22
100 g, Each
Thermo Scientific Chemicals

Adamantane-1-carboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12959.22
100 g, Each
Quantity
Catalog number: A12959.22
also known as A12959-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
828-51-3
IUPAC Name
adamantane-1-carboxylic acid
Molecular Formula
C11H16O2
InChI Key
JIMXXGFJRDUSRO-UHFFFAOYSA-N
SMILES
OC(=O)C12CC3CC(CC(C3)C1)C2
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥98.5%
Water Content (Karl Fischer Titration)
≤0.5%
Appearance (Color)
White to cream or pale yellow
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

It can be used as a stabilizer in the synthesis of monodisperse, highly crystalline CoPt3 nanoparticles1 and porous platinum nanoparticles. It is a potent and reversible CerK inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as a stabilizer in the synthesis of monodisperse, highly crystalline CoPt3 nanoparticles1 and porous platinum nanoparticles. It is a potent and reversible CerK inhibitor.

Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in ethanol, chloroform, and dichloromethane.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text