Thermo Scientific Chemicals

Ethyl isobutyrate, 98%, Thermo Scientific Chemicals

Catalog number: A12966.AE
100 mL, Each
Thermo Scientific Chemicals

Ethyl isobutyrate, 98%, Thermo Scientific Chemicals

Catalog number: A12966.AE
100 mL, Each
Quantity
Catalog number: A12966.AE
also known as A12966-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-62-1
IUPAC Name
ethyl 2-methylpropanoate
Molecular Formula
C6H12O2
InChI Key
WDAXFOBOLVPGLV-UHFFFAOYSA-N
SMILES
CCOC(=O)C(C)C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.3860-1.3905 @ 20?C

Description

It is employed to make flavoring compounds and essences, as a flavoring agent and to flavor animal feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed to make flavoring compounds and essences, as a flavoring agent and to flavor animal feeds.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text