It is used in the synthesis of porous materials by 2-nitroresorcinol/cyanuric chloride thermal polycondensation in emulsions. It is used as an active pharmaceutical intermediate and an intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis of porous materials by 2-nitroresorcinol/cyanuric chloride thermal polycondensation in emulsions. It is used as an active pharmaceutical intermediate and an intermediate for organic synthesis.
Solubility
Slightly soluble in water. Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Konstantin B. Borisenko and Istvan Hargittai. Intramolecular hydrogen bonding and molecular structure of 2-nitroresorcinol from gas-phase electron diffraction.J. Phys. Chem.,1993,97(16),4080-4084.
Charles W. Bock and István Hargittai. Geometrical consequences of resonance-assisted intramolecular hydrogen-bond formation from Ab initio MO calculations on 2-nitroresorcinol.Struct. Chem.,1994,5(5), 307-312.