Thermo Scientific Chemicals

Thiobenzamide, 98%, Thermo Scientific Chemicals

Catalog number: A12973.14
25 g, Each
Thermo Scientific Chemicals

Thiobenzamide, 98%, Thermo Scientific Chemicals

Catalog number: A12973.14
25 g, Each
Quantity
Catalog number: A12973.14
also known as A12973-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2227-79-4
IUPAC Name
benzenecarbothioamide
Molecular Formula
C7H7NS
InChI Key
QIOZLISABUUKJY-UHFFFAOYSA-N
SMILES
NC(=S)C1=CC=CC=C1
Appearance (Color)
Pale yellow to yellow to green
Form
Powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
113.0-121.0?C

Description

Thiobenzamide is useful for the preparation of amide and amidine adducts. It is also used to prepare 4-oxo-4H-chromene-3-carbothioic acid N-phenylamides. Further, it acts as a raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiobenzamide is useful for the preparation of amide and amidine adducts. It is also used to prepare 4-oxo-4H-chromene-3-carbothioic acid N-phenylamides. Further, it acts as a raw material in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text