Thermo Scientific Chemicals

1,2-Dimethoxyethane, 99+%, stab. with BHT, Thermo Scientific Chemicals

Catalog number: A12986.AP
500 mL, Each
Thermo Scientific Chemicals

1,2-Dimethoxyethane, 99+%, stab. with BHT, Thermo Scientific Chemicals

Catalog number: A12986.AP
500 mL, Each
Quantity
Catalog number: A12986.AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
110-71-4
IUPAC Name
1,2-dimethoxyethane
Molecular Formula
C4H10O2
InChI Key
XTHFKEDIFFGKHM-UHFFFAOYSA-N
SMILES
COCCOC
Water Content (Karl Fischer Titration)
≤0.1%
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥99.0%
Identification (FTIR)
Conforms

Description

1,2-Dimethoxyethane is widely used as a solvent for electrolyte of lithium batteries, polysilicones, oligo- and polysaccharides. It plays an important role in Grignard reactions, Suzuki reactions and Stille couplings in organometallic chemistry and in pharmaceutical synthesis. It is a higher boiling point solvent and is used as an alternative to diethyl ether and tetrahydrofuran. It is used for the etching of synthetic polymers like polytetrafluoroethylene and other fluoropolymers with alkali metal dispersions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dimethoxyethane is widely used as a solvent for electrolyte of lithium batteries, polysilicones, oligo- and polysaccharides. It plays an important role in Grignard reactions, Suzuki reactions and Stille couplings in organometallic chemistry and in pharmaceutical synthesis. It is a higher boiling point solvent and is used as an alternative to diethyl ether and tetrahydrofuran. It is used for the etching of synthetic polymers like polytetrafluoroethylene and other fluoropolymers with alkali metal dispersions.

Solubility
Miscible with water, methanol, ethanol, diethyl ether, acetone, tetrahydrofuran, benzene and toluene.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. It can be oxidized on air to form peroxides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text