Thermo Scientific Chemicals

beta-D-Maltose octaacetate, 98%, Thermo Scientific Chemicals

Catalog number: A12988.14
25 g, Each
Thermo Scientific Chemicals

beta-D-Maltose octaacetate, 98%, Thermo Scientific Chemicals

Catalog number: A12988.14
25 g, Each
Quantity
Catalog number: A12988.14
also known as A12988-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22352-19-8
IUPAC Name
[4,5,6-tris(acetyloxy)-3-{[3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate
Molecular Formula
C28H38O19
InChI Key
WOTQVEKSRLZRSX-UHFFFAOYNA-N
SMILES
CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Appearance (Color)
White
Form
Crystalline powder or powder
Assay (unspecified)
≥97.5%
Optical Rotation
+62.5° ± 2.0° (c=1, chloroform)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text