Thermo Scientific Chemicals

trans-4-Nitrocinnamic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12990.14
25 g, Each
Thermo Scientific Chemicals

trans-4-Nitrocinnamic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12990.14
25 g, Each
Quantity
Catalog number: A12990.14
also known as A12990-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
882-06-4
IUPAC Name
(2E)-3-(4-nitrophenyl)prop-2-enoic acid
Molecular Formula
C9H7NO4
InChI Key
XMMRNCHTDONGRJ-ZZXKWVIFSA-N
SMILES
OC(=O)C=CC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
White to cream to yellow
Form
Powder or crystalline powder
Assay from Supplier's CofA
≥98.0%

Description

trans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text