Thermo Scientific Chemicals

2,3-Diaminonaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A12993.06
5 g, Each
Thermo Scientific Chemicals

2,3-Diaminonaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A12993.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A12993.06
also known as A12993-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
771-97-1
IUPAC Name
naphthalene-2,3-diamine
Molecular Formula
C10H10N2
InChI Key
XTBLDMQMUSHDEN-UHFFFAOYSA-N
SMILES
NC1=C(N)C=C2C=CC=CC2=C1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Green to brown
Assay from Suppliers CofA
≥96.0% (HPLC)
Melting Point (clear melt)
190.0-199.0°C

Description

2,3 Diaminonaphthalene is a highly selective colorimetric and fluorometric reagent for selenium detection and also used for the fluorometric determination of nitrite. It is also useful for detection of ketones and alpha-keto acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3 Diaminonaphthalene is a highly selective colorimetric and fluorometric reagent for selenium detection and also used for the fluorometric determination of nitrite. It is also useful for detection of ketones and alpha-keto acids.

Solubility
Slightly soluble in water. Soluble in pyridine, dimethylsulfoxide, dimethyformamide, dil.hydrochloric acid, ethanol, ether and hot methanol.

Notes
Incompatible with oxidizing agents and acids.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text