Thermo Scientific Chemicals

Anisole, 99%, Thermo Scientific Chemicals

Catalog number: A12997.36
500 g, Each
Thermo Scientific Chemicals

Anisole, 99%, Thermo Scientific Chemicals

Catalog number: A12997.36
500 g, Each
Quantity
Catalog number: A12997.36
also known as A12997-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-66-3
IUPAC Name
anisole
Molecular Formula
C7H8O
InChI Key
RDOXTESZEPMUJZ-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5% (UK Source Material)
Assay from Supplier's CofA
≥98.5% (US Sourced Material)
Refractive Index
1.5155-1.5185 @ 20?C
Form
Liquid

Description

Anisole is widely used as a solvent for the synthesis of various organic compounds, anethole, nonylphenol isomer 4-(3',6'-dimethyl-3-heptyl)phenol, perfumes, insect pheromones and pharmaceuticals. It finds application in the preparation of inorganic complexes and materials such as tin-core/tin oxide nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anisole is widely used as a solvent for the synthesis of various organic compounds, anethole, nonylphenol isomer 4-(3′,6′-dimethyl-3-heptyl)phenol, perfumes, insect pheromones and pharmaceuticals. It finds application in the preparation of inorganic complexes and materials such as tin-core/tin oxide nanoparticles.

Solubility
Miscible with methanol and acetone. Immiscible with water.

Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text