2,3-Dibromothiophene, 98+%, Thermo Scientific Chemicals
2,3-Dibromothiophene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dibromothiophene, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12998.06
also known as A12998-06
Price (USD)/ Each
77.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
77.30
Add to cart
2,3-Dibromothiophene, 98+%, Thermo Scientific Chemicals
Catalog numberA12998.06
Price (USD)/ Each
77.30
-
Add to cart
Chemical Identifiers
CAS3140-93-0
IUPAC Name2,3-dibromothiophene
Molecular FormulaC4H2Br2S
InChI KeyATRJNSFQBYKFSM-UHFFFAOYSA-N
SMILESBrC1=C(Br)C=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
Refractive Index1.6295-1.6335 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only