Thermo Scientific Chemicals

Sulfanilamide, 98%, Thermo Scientific Chemicals

Catalog number: A13001.36
500 g, Each
Thermo Scientific Chemicals

Sulfanilamide, 98%, Thermo Scientific Chemicals

Catalog number: A13001.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A13001.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63-74-1
IUPAC Name
4-aminobenzene-1-sulfonamide
Molecular Formula
C6H8N2O2S
InChI Key
FDDDEECHVMSUSB-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)S(N)(=O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
162.0-168.0?C

Description

Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.

Solubility
Soluble in water, acetone, ethanol, glycerol, propylene glycol and hydrochloric acid. Insoluble in chloroform, ether, benzene and petroleum ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text