Thermo Scientific Chemicals

Benzyl carbazate, 98+%, Thermo Scientific Chemicals

Catalog number: A13005.06
5 g, Each
Thermo Scientific Chemicals

Benzyl carbazate, 98+%, Thermo Scientific Chemicals

Catalog number: A13005.06
5 g, Each
Quantity
Catalog number: A13005.06
also known as A13005-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5331-43-1
IUPAC Name
(benzyloxy)carbohydrazide
Molecular Formula
C8H10N2O2
InChI Key
RXUBZLMIGSAPEJ-UHFFFAOYSA-N
SMILES
NNC(=O)OCC1=CC=CC=C1
Appearance (Color)
White to pale yellow
Form
Crystalline powder or flakes
Assay (GC)
>98.0%

Description

Benzyl carbazate is used as a reagent for preparation of benzyloxycarbonyl (Z, Cbz) hydrazides of carboxylic acids, particularly N-protected amino acids. Subsequent hydrogenolysis gives the hydrazide which can be converted to the azide for coupling with a further amino acid residue by the azide method. It is also used in the preparation of simple acyl or sulfonyl hydrazides avoiding diacylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl carbazate is used as a reagent for preparation of benzyloxycarbonyl (Z, Cbz) hydrazides of carboxylic acids, particularly N-protected amino acids. Subsequent hydrogenolysis gives the hydrazide which can be converted to the azide for coupling with a further amino acid residue by the azide method. It is also used in the preparation of simple acyl or sulfonyl hydrazides avoiding diacylation.

Solubility
Soluble in water (slightly), methanol and DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle under inert gas. Protect from moisture. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text