Thermo Scientific Chemicals

5-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13010.22
100 g, Each
Thermo Scientific Chemicals

5-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13010.22
100 g, Each
Quantity
Catalog number: A13010.22
also known as A13010-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-97-9
IUPAC Name
2-hydroxy-5-nitrobenzoic acid
Molecular Formula
C7H5NO5
InChI Key
PPDRLQLKHRZIJC-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=CC=C1O)[N+]([O-])=O
Appearance (Color)
Pale brown
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms

Description

5-Nitrosalicylic acid is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrosalicylic acid is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic, pharmaceutical intermediate.

Solubility
Soluble in water 1g/1475ml .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text