Thermo Scientific Chemicals

1-Bromo-4-ethylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13013.09
10 g, Each
Thermo Scientific Chemicals

1-Bromo-4-ethylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13013.09
10 g, Each
Quantity
Catalog number: A13013.09
also known as A13013-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1585-07-5
IUPAC Name
1-bromo-4-ethylbenzene
Molecular Formula
C8H9Br
InChI Key
URFPRAHGGBYNPW-UHFFFAOYSA-N
SMILES
CCC1=CC=C(Br)C=C1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5435-1.5465 @ 20?C

Description

1-Bromo-4-ethylbenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-ethylbenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Avoid excessive heat and light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text