Thermo Scientific Chemicals

4-Methoxybenzhydrazide, 98+%, Thermo Scientific Chemicals

Catalog number: A13015.30
250 g, Each
Thermo Scientific Chemicals

4-Methoxybenzhydrazide, 98+%, Thermo Scientific Chemicals

Catalog number: A13015.30
250 g, Each
Quantity
Catalog number: A13015.30
also known as A13015-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3290-99-1
IUPAC Name
4-methoxybenzohydrazide
Molecular Formula
C8H10N2O2
InChI Key
REKQLYUAUXYJSZ-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)C(=O)NN
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Melting Point (clear melt)
134.0-140.0?C
Appearance (Color)
Pale cream

Description

4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text