Thermo Scientific Chemicals

Sodium fluoride, 99%, Thermo Scientific Chemicals

Catalog number: A13019.30
250 g, Each
Thermo Scientific Chemicals

Sodium fluoride, 99%, Thermo Scientific Chemicals

Catalog number: A13019.30
250 g, Each
Quantity
Catalog number: A13019.30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7681-49-4
IUPAC Name
sodium fluoride
Molecular Formula
FNa
InChI Key
PUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES
[F-].[Na+]
Appearance (Color)
White
Form
Powder
Assay from Supplier's CofA
≥98.5 to ≤101.5%
Loss on Drying
<0.5% (150?C/constant wt)

Description

Sodium fluoride is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium fluoride is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

Solubility
Slightly soluble in water. Insoluble in alcohol

Notes
It must be kept away from oxidizing agents, metals, acids, alkalis. It is less hygroscopic.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text