2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acid chlorides, acid anhydrides, oxidizing agents.
RUO – Research Use Only
General References:
- Y Chen.; C Qian.; J Sun. Fluoro-substituted 2, 6-bis (imino) pyridyl iron and cobalt complexes: high-activity ethylene oligomerization catalysts. Organometallics. 200322 (6), 1231-1236.
- CN Chen.; LL Lv.; FQ Ji.; Q Chen.; H Xu.; CW Niu. Design and synthesis of N-2, 6-difluorophenyl-5-methoxyl-1, 2, 4-triazolo [1, 5-a]-pyrimidine-2-sulfonamide as acetohydroxyacid synthase inhibitor. Bioorganic & medicinal chemistry. 200917 (8), 3011-3017.