Thermo Scientific Chemicals

2,6-Difluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A13020.22
100 g, Each
Thermo Scientific Chemicals

2,6-Difluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A13020.22
100 g, Each
Quantity
Catalog number: A13020.22
also known as A13020-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5509-65-9
IUPAC Name
2,6-difluoroaniline
Molecular Formula
C6H5F2N
InChI Key
ODUZJBKKYBQIBX-UHFFFAOYSA-N
SMILES
NC1=C(F)C=CC=C1F
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.5060-1.5100 @ 20?C

Description

2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acid chlorides, acid anhydrides, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text