Thermo Scientific Chemicals

2-Aminoisobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: A13021.22
100 g, Each
Thermo Scientific Chemicals

2-Aminoisobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: A13021.22
100 g, Each
Quantity
Catalog number: A13021.22
also known as A13021-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62-57-7
IUPAC Name
2-amino-2-methylpropanoic acid
Molecular Formula
C4H9NO2
InChI Key
FUOOLUPWFVMBKG-UHFFFAOYSA-N
SMILES
CC(C)(N)C(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or lumps
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (Silylated GC)
≥98.5%

Description

2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.

Solubility
Soluble in water. Insoluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text