Thermo Scientific Chemicals

3-Phenyl-1-propanol, 99%, Thermo Scientific Chemicals

Catalog number: A13022.30
250 g, Each
Thermo Scientific Chemicals

3-Phenyl-1-propanol, 99%, Thermo Scientific Chemicals

Catalog number: A13022.30
250 g, Each
Quantity
Catalog number: A13022.30
also known as A13022-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-97-4
IUPAC Name
3-phenylpropan-1-ol
Molecular Formula
C9H12O
InChI Key
VAJVDSVGBWFCLW-UHFFFAOYSA-N
SMILES
OCCCC1=CC=CC=C1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5255-1.5285 @ 20?C

Description

Ester derivatives are used as fragrance component in fresh flower compositions, like lilac, hyacinth and lily of the valley because of the balsamic odor character. It is also a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenyl-1-propanol has a natural fragrance with antimicrobial properties against bacteria and molds. It is used in combination with Heliotropin or Piperonal as a preservative for cosmetic products. Its ester derivatives are used as fragrance component in fresh flower compositions, like lilac, hyacinth and lily of the valley because of the balsamic odor character. It is also a useful biochemical for proteomics research.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text