Thermo Scientific Chemicals

Aniline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A13024.36
500 g, Each
Thermo Scientific Chemicals

Aniline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A13024.36
500 g, Each
Quantity
Catalog number: A13024.36
also known as A13024-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-04-1
IUPAC Name
hydrogen aniline chloride
Molecular Formula
C6H8ClN
InChI Key
MMCPOSDMTGQNKG-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1=CC=CC=C1
Appearance (Color)
White to grey or pale yellow or pale green
Form
Crystals or crystalline powder or powder or flakes
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Melting Point (clear melt)
195-201?C
Identification (FTIR)
Conforms

Description

Aniline hydrochloride was used in the preparation of polyaniline coated poly(styrene-co-styrene sulfonate) nanoparticles. It was used to study the induction of Nei-like DNA glycosylases (NEIL1/2)-mediated base excision repair(BER) in rat spleen and 8-oxoguanine glycosylase 1-mediated BER due to aniline exposure. It is used in printing inks, clothing marking inks, paints and paint removers. It is also used as a stove polisher and shoe polisher, as well as in the synthesis of dyes, crayons and antioxidants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aniline hydrochloride was used in the preparation of polyaniline coated poly(styrene-co-styrene sulfonate) nanoparticles. It was used to study the induction of Nei-like DNA glycosylases (NEIL1/2)-mediated base excision repair(BER) in rat spleen and 8-oxoguanine glycosylase 1-mediated BER due to aniline exposure. It is used in printing inks, clothing marking inks, paints and paint removers. It is also used as a stove polisher and shoe polisher, as well as in the synthesis of dyes, crayons and antioxidants.

Solubility
Soluble in water (1070 g/L).

Notes
It is light sensitive and hygroscopic. Incompatible with oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text